Assay Details
Gene Symbol: PIMREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005693431.3 | XP_005693488.1 | 4 - 5 | 790 | 87 | |
GenBank mRNA | KA333978.1 | - | 2 - 3 | 262 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
