Assay Details
| Gene Symbol: TRAPPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005693537.2 | XP_005693594.1 | 3 - 4 | 233 | 84 | |
| GenBank mRNA | JO643802.2 | - | 2 - 3 | 571 | 84 | |
More Information
Set Membership: |
|

TRAPPC1
Genomic Map