Assay Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005694736.3 | XP_005694793.1 | 3 - 4 | 1747 | 66 | |
| XM_013972799.2 | XP_013828253.1 | 2 - 3 | 419 | 66 | ||
More Information
Set Membership: |
|

TMEM267
Genomic Map