Assay Details
Gene Symbol: DEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039020.1 | XP_017894509.1 | 4 - 5 | 738 | 75 | |
XM_018039021.1 | XP_017894510.1 | 4 - 5 | 496 | 75 | ||
XM_018039022.1 | XP_017894511.1 | 4 - 5 | 738 | 75 | ||
GenBank mRNA | JO582180.1 | - | 2323 | 75 | ||
JO582184.1 | - | 2326 | 75 | |||
JR126897.1 | - | 4661 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
