Assay Details
Gene Symbol: NDUFB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040705.1 | XP_017896194.1 | 3 - 4 | 566 | 76 | |
GenBank mRNA | JO447006.2 | - | 3 - 4 | 595 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
