Assay Details
| Gene Symbol: ARPC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040844.1 | XP_017896333.1 | 7 - 8 | 965 | 79 | |
| GenBank mRNA | JO645353.2 | - | 7 - 8 | 960 | 79 | |
More Information
Set Membership: |
|

ARPC1A
Genomic Map