Assay Details
Gene Symbol: TLR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041932.1 | XP_017897421.1 | 2 - 3 | 792 | 92 | |
XM_018041934.1 | XP_017897423.1 | 2 - 3 | 782 | 92 | ||
XM_018041935.1 | XP_017897424.1 | 2 - 3 | 791 | 92 | ||
GenBank mRNA | GU936194.1 | - | 1 - 2 | 436 | 92 | |
GU936195.1 | - | 1 - 2 | 436 | 92 | ||
GU936196.1 | - | 1 - 2 | 436 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
