Assay Details
Gene Symbol: NKIRAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041841.1 | XP_017897330.1 | 2 - 3 | 217 | 78 | |
GenBank mRNA | JO621310.2 | - | 2 - 3 | 760 | 78 | |
JR131441.1 | - | 1728 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
