Assay Details
| Gene Symbol: ATP6V0E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963581.2 | XP_013819035.2 | 1 - 2 | 321 | 82 | |
| GenBank mRNA | JO631957.2 | - | 3 - 4 | 1659 | 82 | |
| JT624308.1 | - | 1 - 2 | 135 | 82 | ||
More Information
Set Membership: |
|

ATP6V0E2
Genomic Map