Assay Details
| Gene Symbol: TMCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060046.1 | XP_017915535.1 | 2 - 3 | 574 | 76 | |
| GenBank mRNA | JO520637.2 | - | 1 - 2 | 110 | 76 | |
More Information
Set Membership: |
|

TMCC2
Genomic Map