Assay Details
| Gene Symbol: DEGS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060204.1 | XP_017915693.1 | 2 - 3 | 967 | 76 | |
| GenBank mRNA | JO562239.2 | - | 1 - 2 | 391 | 76 | |
| JO630306.2 | - | 1 - 2 | 395 | 76 | ||
More Information
Set Membership: |
|

DEGS1
Genomic Map