Assay Details
| Gene Symbol: TMEM9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060839.1 | XP_017916328.1 | 3 - 4 | 281 | 63 | |
| XM_018060840.1 | XP_017916329.1 | 2 - 3 | 568 | 63 | ||
| GenBank mRNA | JO433782.2 | - | 2 - 3 | 212 | 63 | |
| JO612478.2 | - | 3 - 4 | 216 | 63 | ||
More Information
Set Membership: |
|

TMEM9
Genomic Map