Assay Details
Gene Symbol: PRSS33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039852.1 | XP_017895341.1 | 4 - 5 | 556 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
