Assay Details
Gene Symbol: TBCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675163.3 | XP_005675220.2 | 4 - 5 | 989 | 88 | |
XM_013963912.2 | XP_013819366.2 | 4 - 5 | 1179 | 88 | ||
XM_013963915.2 | XP_013819369.2 | 4 - 5 | 990 | 88 | ||
GenBank mRNA | JO595287.2 | - | 4 - 5 | 1060 | 88 | |
JT706010.1 | - | 5 - 6 | 1119 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
