Assay Details
| Gene Symbol: CASP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676364.3 | XP_005676421.1 | 5 - 6 | 971 | 75 | |
| XM_005676365.3 | XP_005676422.1 | 3 - 4 | 847 | 75 | ||
| XM_013971078.2 | XP_013826532.1 | 4 - 5 | 909 | 75 | ||
| XM_018060934.1 | XP_017916423.1 | 5 - 6 | 971 | 75 | ||
| GenBank mRNA | JO578360.1 | - | 6 - 7 | 1681 | 75 | |
| JQ342087.1 | - | 2 - 3 | 347 | 75 | ||
More Information
Set Membership: |
|

CASP8
Genomic Map