Assay Details
| Gene Symbol: ICOS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005676405.3 | XP_005676462.1 | 4 - 5 | 639 | 74 | |
| GenBank mRNA | JO336941.2 | - | 2 - 3 | 179 | 74 | |
| JO591812.2 | - | 4 - 5 | 637 | 74 | ||
More Information
Set Membership: |
|

ICOS
Genomic Map