Assay Details
| Gene Symbol: RAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680676.3 | XP_005680733.1 | 4 - 5 | 498 | 69 | |
| GenBank mRNA | JO579349.1 | - | 3 - 4 | 2538 | 69 | |
| JP350369.2 | - | 4 - 5 | 347 | 69 | ||
More Information
Set Membership: |
|

RAC2
Genomic Map