Assay Details
Gene Symbol: ECM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005684160.3 | XP_005684217.1 | 9 - 10 | 2099 | 73 | |
XM_005684161.3 | XP_005684218.1 | 9 - 10 | 2033 | 73 | ||
XM_013966189.2 | XP_013821643.1 | 9 - 10 | 2103 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
