Assay Details
Gene Symbol: CIAO2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685668.3 | XP_005685725.1 | 2 - 3 | 511 | 73 | |
GenBank mRNA | JO439811.2 | - | 3 - 4 | 570 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
