Assay Details
| Gene Symbol: PIGH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685998.3 | XP_005686055.2 | 1 - 2 | 355 | 56 | |
| XM_013967247.2 | XP_013822701.1 | 1 - 2 | 366 | 56 | ||
| XM_013967248.2 | XP_013822702.1 | 1 - 2 | 369 | 56 | ||
| GenBank mRNA | JO438303.2 | - | 1 - 2 | 271 | 56 | |
| KA315168.1 | - | 3 - 4 | 945 | 56 | ||
More Information
Set Membership: |
|

PIGH
Genomic Map