Assay Details
Gene Symbol: OSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005689204.2 | XP_005689261.1 | 2 - 3 | 1321 | 69 | |
XM_005689206.2 | XP_005689263.1 | 2 - 3 | 1321 | 69 | ||
GenBank mRNA | JO436600.2 | - | 2 - 3 | 1151 | 69 | |
KA335045.1 | - | 2 - 3 | 553 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
