Assay Details
| Gene Symbol: DUSP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690488.3 | XP_005690545.1 | 3 - 4 | 1426 | 61 | |
| GenBank mRNA | KA337429.1 | - | 1 - 2 | 330 | 61 | |
More Information
Set Membership: |
|

DUSP10
Genomic Map