Assay Details
Gene Symbol: FAM186B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963823.2 | XP_013819277.2 | 4 - 5 | 2203 | 66 | |
XM_018047835.1 | XP_017903324.1 | 3 - 4 | 2994 | 66 | ||
XM_018047836.1 | XP_017903325.1 | 5 - 6 | 2361 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
