Assay Details
| Gene Symbol: LOC102180747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964712.2 | XP_013820166.2 | 4 - 5 | 833 | 78 | |
| XM_018049561.1 | XP_017905050.1 | 4 - 5 | 837 | 78 | ||
| XM_018049562.1 | XP_017905051.1 | 4 - 5 | 534 | 78 | ||
More Information
Set Membership: |
|

LOC102180747
Genomic Map