Assay Details
Gene Symbol: RAB6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051101.1 | XP_017906590.1 | 3 - 4 | 610 | 87 | |
GenBank mRNA | JO634061.2 | - | 5 - 6 | 686 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
