Assay Details
Gene Symbol: LOC102171556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061126.1 | XP_017916615.1 | 21 - 22 | 2460 | 75 | |
GenBank mRNA | KA337612.1 | - | 14 - 15 | 1924 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
