Assay Details
| Gene Symbol: RAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065431.1 | XP_017920920.1 | 3 - 4 | 493 | 56 | |
| XM_018065432.1 | XP_017920921.1 | 3 - 4 | 1208 | 56 | ||
| XM_018065433.1 | XP_017920922.1 | 3 - 4 | 741 | 56 | ||
More Information
Set Membership: |
|

RAC3
Genomic Map