Assay Details
Gene Symbol: MOCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972777.2 | XP_013828231.2 | 5 - 6 | 606 | 85 | |
XM_018065594.1 | XP_017921083.1 | 4 - 5 | 495 | 85 | ||
XM_018065595.1 | XP_017921084.1 | 3 - 4 | 1775 | 85 | ||
XM_018065596.1 | XP_017921085.1 | 3 - 4 | 431 | 85 | ||
GenBank mRNA | JO436993.2 | - | 5 - 6 | 678 | 85 | |
JO439117.2 | - | 4 - 5 | 533 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
