Assay Details
| Gene Symbol: PIGV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974043.2 | XP_013829497.1 | 3 - 4 | 1090 | 63 | |
| XM_013974055.2 | XP_013829509.1 | 2 - 3 | 1223 | 63 | ||
| XM_018056505.1 | XP_017911994.1 | 3 - 4 | 289 | 63 | ||
| XM_018056509.1 | XP_017911998.1 | 3 - 4 | 260 | 63 | ||
| GenBank mRNA | JT672247.1 | - | 2 - 3 | 1679 | 63 | |
More Information
Set Membership: |
|

PIGV
Genomic Map