Assay Details
Gene Symbol: LOC102186356 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677150.3 | XP_005677207.2 | 5 - 6 | 876 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
