Assay Details
Gene Symbol: DELE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683338.3 | XP_005683395.2 | 5 - 6 | 1482 | 60 | |
XM_018050215.1 | XP_017905704.1 | 5 - 6 | 1482 | 60 | ||
GenBank mRNA | JT659818.1 | - | 6 - 7 | 750 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
