Assay Details
| Gene Symbol: MFSD4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684568.3 | XP_005684625.1 | 1 - 2 | 212 | 55 | |
| GenBank mRNA | JO625453.2 | - | 1 - 2 | 372 | 55 | |
More Information
Set Membership: |
|

MFSD4B
Genomic Map