Assay Details
Gene Symbol: PPP1R36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685974.3 | XP_005686031.1 | 4 - 5 | 357 | 63 | |
XM_005685976.3 | XP_005686033.1 | 3 - 4 | 270 | 63 | ||
XM_005685977.3 | XP_005686034.1 | 4 - 5 | 357 | 63 | ||
XM_005685978.3 | XP_005686035.1 | 2 - 3 | 213 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
