Assay Details
| Gene Symbol: THNSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686693.3 | XP_005686750.3 | 7 - 8 | 1210 | 65 | |
| XM_005686694.3 | XP_005686751.3 | 6 - 7 | 979 | 65 | ||
More Information
Set Membership: |
|

THNSL2
Genomic Map