Assay Details
| Gene Symbol: IMPAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688998.3 | XP_005689055.2 | 2 - 3 | 810 | 80 | |
| GenBank mRNA | JT763441.1 | - | 3 - 4 | 892 | 80 | |
More Information
Set Membership: |
|

IMPAD1
Genomic Map