Assay Details
| Gene Symbol: CPTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060542.1 | XP_017916031.1 | 2 - 3 | 236 | 79 | |
| XM_018060543.1 | XP_017916032.1 | 2 - 3 | 231 | 79 | ||
| GenBank mRNA | KA351916.1 | - | 2 - 3 | 255 | 79 | |
More Information
Set Membership: |
|

CPTP
Genomic Map