Assay Details
Gene Symbol: POP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692232.3 | XP_005692289.1 | 4 - 5 | 593 | 66 | |
GenBank mRNA | JO610184.2 | - | 3 - 4 | 679 | 66 | |
JO629273.2 | - | 2 - 3 | 84 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
