Assay Details
| Gene Symbol: ALDH3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064739.1 | XP_017920228.1 | 10 - 11 | 1523 | 67 | |
| GenBank mRNA | JO427893.1 | - | 5 - 6 | 1023 | 67 | |
| JO428215.1 | - | 1 - 2 | 1023 | 67 | ||
More Information
Set Membership: |
|

ALDH3A2
Genomic Map