Assay Details
Gene Symbol: MS4A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042748.1 | XP_017898237.1 | 4 - 5 | 694 | 100 | |
GenBank mRNA | JT668557.1 | - | 2 - 3 | 178 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
