Assay Details
| Gene Symbol: PHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048523.1 | XP_017904012.1 | 8 - 9 | 2103 | 60 | |
| XM_018048524.1 | XP_017904013.1 | 8 - 9 | 2079 | 60 | ||
| XM_018048525.1 | XP_017904014.1 | 7 - 8 | 1929 | 60 | ||
More Information
Set Membership: |
|

PHC1
Genomic Map