Assay Details
| Gene Symbol: ARID3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052839.1 | XP_017908328.1 | 5 - 6 | 1148 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARID3C| Gene Symbol: ARID3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052839.1 | XP_017908328.1 | 5 - 6 | 1148 | 63 | |
Set Membership: |
|