Assay Details
Gene Symbol: HERPUD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971020.2 | XP_013826474.1 | 5 - 6 | 1157 | 60 | |
XM_018062025.1 | XP_017917514.1 | 5 - 6 | 678 | 60 | ||
GenBank mRNA | JO319972.2 | - | 5 - 6 | 569 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
