Assay Details
| Gene Symbol: RNF138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039774.1 | XP_017895263.1 | 4 - 5 | 567 | 95 | |
| XM_018039775.1 | XP_017895264.1 | 4 - 5 | 567 | 95 | ||
| GenBank mRNA | JT728362.1 | - | 5 - 6 | 500 | 95 | |
More Information
Set Membership: |
|

RNF138
Genomic Map