Assay Details
Gene Symbol: MICOS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054827.1 | XP_017910316.1 | 3 - 4 | 873 | 64 | |
GenBank mRNA | JO574776.2 | - | 3 - 4 | 286 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
