Assay Details
Gene Symbol: TSC22D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040776.1 | XP_017896265.1 | 4 - 5 | 2004 | 69 | |
GenBank mRNA | JO392910.2 | - | 1 - 2 | 587 | 69 | |
JO427808.1 | - | 1 - 2 | 659 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
