Assay Details
Gene Symbol: CLCA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678150.2 | XP_005678207.2 | 2 - 3 | 320 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
