Assay Details
| Gene Symbol: GNG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018045676.1 | XP_017901165.1 | 2 - 3 | 843 | 128 | |
| GenBank mRNA | JO542761.2 | - | 2 - 3 | 402 | 128 | |
More Information
Set Membership: |
|

GNG5
Genomic Map