Assay Details
Gene Symbol: POU6F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963798.2 | XP_013819252.1 | 9 - 10 | 2876 | 57 | |
XM_018047756.1 | XP_017903245.1 | 10 - 11 | 1819 | 57 | ||
XM_018047757.1 | XP_017903246.1 | 9 - 10 | 2836 | 57 | ||
XM_018047758.1 | XP_017903247.1 | 10 - 11 | 1816 | 57 | ||
XM_018047759.1 | XP_017903248.1 | 10 - 11 | 1730 | 57 | ||
GenBank mRNA | JO617667.2 | - | 1 - 2 | 865 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
