Assay Details
Gene Symbol: CLEC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680852.3 | XP_005680909.2 | 1 - 2 | 206 | 65 | |
XM_013964211.2 | XP_013819665.2 | 1 - 2 | 206 | 65 | ||
XM_018048489.1 | XP_017903978.1 | 1 - 2 | 206 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
