Assay Details
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005681330.3 | XP_005681387.2 | 2 - 3 | 426 | 68 | |
XM_018049236.1 | XP_017904725.1 | 2 - 3 | 464 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
